accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 82010-31-9

(S)HNOHOCH3CH3CH3OH3CCH3

CAS RN 82010-31-9

IUPAC Name: tert-butyl N-[(2S)-1-hydroxy-4-methylpentan-2-yl]carbamate
Synonyms: tert-butyl N-[(2S)-1-hydroxy-4-methylpentan-2-yl]carbamate
Molecular Weight (g/mol): 217.31
Molecular Formula: C11H23NO3
InChi Key: LQTMEOSBXTVYRM-VIFPVBQESA-N
SMILES: CC(C)C[C@@H](CO)NC(=O)OC(C)(C)C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)