accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 819849-20-2

HOBOHFNOO

CAS RN 819849-20-2

IUPAC Name: (2-fluoro-5-nitrophenyl)boronic acid
Synonyms: 2-fluoro-5-nitrophenylboronic acid
Molecular Weight (g/mol): 184.92
Molecular Formula: C6H5BFNO4
InChi Key: SYAFEPQKPQIRAL-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results