accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 819-83-0

NaNaOPOOOHOHO

CAS RN 819-83-0

IUPAC Name: tetrasodium 3-(phosphonatooxy)pentane-1,5-diol 3-(phosphonatooxy)propane-1,2-diol
Synonyms: tetrasodium 3-(phosphonatooxy)pentane-1,5-diol DL-glycerol 3-phosphate
Molecular Weight (g/mol): 460.13
Molecular Formula: C8H18Na4O12P2
InChi Key: WENPEHSLKZUZKX-UHFFFAOYNA-J
SMILES: [Na+].[Na+].[Na+].[Na+].OCC(O)COP([O-])([O-])=O.OCCC(CCO)OP([O-])([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results