accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 81759-25-3

NON

CAS RN 81759-25-3

IUPAC Name: 1-(pyrrolidine-1-carbonyl)pyrrolidine
Synonyms: 1-(pyrrolidine-1-carbonyl)pyrrolidine
Molecular Weight (g/mol): 168.24
Molecular Formula: C9H16N2O
InChi Key: HPTQKSXAQBHFKL-UHFFFAOYSA-N
SMILES: O=C(N1CCCC1)N1CCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results