accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NSOHOO

CAS RN 81-16-3

IUPAC Name: 2-aminonaphthalene-1-sulfonic acid
Synonyms: tobias acid
Molecular Weight (g/mol): 223.25
Molecular Formula: C10H9NO3S
InChi Key: GWIAAIUASRVOIA-UHFFFAOYSA-N
SMILES: NC1=CC=C2C=CC=CC2=C1S(O)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)