CAS RN 80913-66-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Methylmorpholine N-oxide hydrate, 97%
CAS: 80913-66-2 Molecular Formula: C5H11NO2 Molecular Weight (g/mol): 117.15 MDL Number: MFCD00005947 InChI Key: LFTLOKWAGJYHHR-UHFFFAOYSA-N Synonym: 4-methylmorpholine n-oxide,4-methylmorpholine 4-oxide,n-methylmorpholine oxide,n-methylmorpholine n-oxide,morpholine, 4-methyl-, 4-oxide,4-methylmorpholine-4-oxide,methyl morpholine oxide,n-methylmorpholine-n-oxide,nmo,unii-arc64pkj0f PubChem CID: 82029 ChEBI: CHEBI:52093 IUPAC Name: 4-methyl-4-oxidomorpholin-4-ium SMILES: C[N+]1([O-])CCOCC1