accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 80745-07-9

CH3SOOClH3CCH3OCH3

CAS RN 80745-07-9

IUPAC Name: 4-methoxy-2,3,6-trimethylbenzene-1-sulfonyl chloride
Synonyms: 4-methoxy-2,3,6-trimethylbenzenesulfonyl chloride
Molecular Weight (g/mol): 248.72
Molecular Formula: C10H13ClO3S
InChi Key: DWLYVEYCCPEHLX-UHFFFAOYSA-N
SMILES: COC1=C(C)C(C)=C(C(C)=C1)S(Cl)(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)