accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 80741-39-5

H3CH3CCH3OONH(S)ClClOO

CAS RN 80741-39-5

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(3,4-dichlorophenyl)propanoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(3,4-dichlorophenyl)propanoate
Molecular Weight (g/mol): 333.19
Molecular Formula: C14H16Cl2NO4
InChi Key: UGZIQCCPEDCGGN-NSHDSACASA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Cl)C(Cl)=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results