accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 80143-73-3

NHFF

CAS RN 80143-73-3

IUPAC Name: 4-fluoro-N-[(4-fluorophenyl)methyl]aniline
Synonyms: 4-fluoro-N-[(4-fluorophenyl)methyl]aniline
Molecular Weight (g/mol): 219.24
Molecular Formula: C13H11F2N
InChi Key: OQVIJNAAQBTAAY-UHFFFAOYSA-N
SMILES: FC1=CC=C(CNC2=CC=C(F)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results