accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 80141-91-9

CH3FHO

CAS RN 80141-91-9

IUPAC Name: (4-fluoro-2-methylphenyl)methanol
Synonyms: (4-fluoro-2-methylphenyl)methanol
Molecular Weight (g/mol): 140.16
Molecular Formula: C8H9FO
InChi Key: YSULUXOLTMBSFF-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC=C1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results