CAS RN 8001-29-4
CAS RN 8001-29-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Cottonseed oil, pure
CAS: 8001-29-4 MDL Number: MFCD00130872
2
Cottonseed Oil, MP Biomedicals
CAS: 8001-29-4