accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 79950-42-8

CH2HOO

CAS RN 79950-42-8

IUPAC Name: 3-hydroxy-2-(prop-2-en-1-yl)benzaldehyde
Synonyms: 3-hydroxy-2-(prop-2-en-1-yl)benzaldehyde
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: QVHRAGBOMUXWRI-UHFFFAOYSA-N
SMILES: OC1=CC=CC(C=O)=C1CC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results