accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOFCl

CAS RN 79944-64-2

IUPAC Name: (3-chloro-5-fluorophenyl)methanol
Synonyms: (3-chloro-5-fluorophenyl)methanol
Molecular Weight (g/mol): 160.57
Molecular Formula: C7H6ClFO
InChi Key: VJTJBAMDTCIMOB-UHFFFAOYSA-N
SMILES: OCC1=CC(F)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results