accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH

CAS RN 79887-10-8

IUPAC Name: 1-ethynyl-4-pentylbenzene
Synonyms: 1-ethynyl-4-pentylbenzene
Molecular Weight (g/mol): 172.27
Molecular Formula: C13H16
InChi Key: APGNXGIUUTWIRE-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(C=C1)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results