accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOOOOCH3

CAS RN 794-94-5

IUPAC Name: 4-methoxybenzoyl 4-methoxybenzoate
Synonyms: p-anisic anhydride
Molecular Weight (g/mol): 286.28
Molecular Formula: C16H14O5
InChi Key: YGMHIBLUWGDWKP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)OC(=O)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results