accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SBrOH

CAS RN 79387-71-6

IUPAC Name: (5-bromothiophen-2-yl)methanol
Synonyms: (5-bromothiophen-2-yl)methanol
Molecular Weight (g/mol): 193.06
Molecular Formula: C5H5BrOS
InChi Key: DBRSLMCLMMFHEF-UHFFFAOYSA-N
SMILES: OCC1=CC=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results