accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 79069-14-0

IUPAC Name: tert-butyl N-(1-hydroxy-3-methylbutan-2-yl)carbamate
Synonyms: tert-butyl N-(1-hydroxy-3-methylbutan-2-yl)carbamate
Molecular Weight (g/mol): 203.28
Molecular Formula: C10H21NO3
InChi Key: OOQRRYDVICNJGC-UHFFFAOYNA-N
SMILES: CC(C)C(CO)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results