accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 79-05-0

IUPAC Name: propanamide
Synonyms: propionamide proprionamide propionimidic acid amid kyseliny propionove propanimidic acid
Molecular Weight (g/mol): 73.1
Molecular Formula: C3H7NO
InChi Key: QLNJFJADRCOGBJ-UHFFFAOYSA-N
SMILES: CCC(N)=O
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results