accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 78985-13-4

CH3H3COHCH3CH3

CAS RN 78985-13-4

IUPAC Name: (2,3,5,6-tetramethylphenyl)methanol
Synonyms: (2,3,5,6-tetramethylphenyl)methanol
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: QVMOEEBACLJUFD-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)C(CO)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results