accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OHNBOHHO

CAS RN 78887-39-5

IUPAC Name: (3-acetamidophenyl)boronic acid
Synonyms: 3-acetamidophenylboronic acid
Molecular Weight (g/mol): 178.98
Molecular Formula: C8H10BNO3
InChi Key: IBTSWKLSEOGJGJ-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results