accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNH2

CAS RN 78881-21-7

IUPAC Name: 4-amino-3-methylbenzonitrile
Synonyms: 4-amino-3-methylbenzonitrile
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: MBZDCUMFFPWLTJ-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results