accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SiH3CCH3Cl

CAS RN 78704-49-1

IUPAC Name: [2-(4-chlorophenyl)ethynyl]trimethylsilane
Synonyms: [2-(4-chlorophenyl)ethynyl]trimethylsilane
Molecular Weight (g/mol): 208.76
Molecular Formula: C11H13ClSi
InChi Key: HVIHXUCWKNRJTC-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results