accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFFFFClO

CAS RN 785-56-8

IUPAC Name: 3,5-bis(trifluoromethyl)benzoyl chloride
Synonyms: 3,5-bis(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 276.56
Molecular Formula: C9H3ClF6O
InChi Key: WAKMMQSMEDJRRI-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(=C1)C(Cl)=O)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results