accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 780-69-8

IUPAC Name: triethoxy(phenyl)silane
Synonyms: silane, triethoxyphenyl-
Molecular Weight (g/mol): 240.37
Molecular Formula: C12H20O3Si
InChi Key: JCVQKRGIASEUKR-UHFFFAOYSA-N
SMILES: CCO[Si](OCC)(OCC)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results