accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NOOH

CAS RN 77940-94-4

IUPAC Name: 3-(piperidin-1-yl)benzoic acid
Synonyms: 3-(piperidin-1-yl)benzoic acid
Molecular Weight (g/mol): 205.26
Molecular Formula: C12H15NO2
InChi Key: FLQRORYAJSTYLT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results