accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7791-23-3

ClSeClO

CAS RN 7791-23-3

IUPAC Name: selenooyl dichloride
Synonyms: selenium oxychloride selenium oxydichloride
Molecular Weight (g/mol): 165.87
Molecular Formula: Cl2OSe
InChi Key: LIRMPIXWGWLNND-UHFFFAOYSA-N
SMILES: Cl[Se](Cl)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)