accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7790-60-5

KKOWOOO

CAS RN 7790-60-5

IUPAC Name: dipotassium dioxotungstenbis(olate)
Synonyms: dipotassium tungstate
Molecular Weight (g/mol): 326.03
Molecular Formula: K2O4W
InChi Key: AAQNGTNRWPXMPB-UHFFFAOYSA-N
SMILES: [K+].[K+].[O-][W]([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)