accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7790-39-8

IIPt

CAS RN 7790-39-8

IUPAC Name: platinum(2+) diiodide
Synonyms: platinum(2+) diiodide
Molecular Weight (g/mol): 448.89
Molecular Formula: I2Pt
InChi Key: ZXDJCKVQKCNWEI-UHFFFAOYSA-L
SMILES: [I-].[I-].[Pt++]
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results