accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7790-38-7

PdII

CAS RN 7790-38-7

IUPAC Name: palladium(2+) diiodide
Synonyms: palladium(II) iodide
Molecular Weight (g/mol): 360.23
Molecular Formula: I2Pd
InChi Key: HNNUTDROYPGBMR-UHFFFAOYSA-L
SMILES: [Pd++].[I-].[I-]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results