CAS RN 7790-33-2
IUPAC Name:
manganese(2+) diiodide
Synonyms:
manganese(II) iodide
Molecular Weight (g/mol):
308.75
Molecular Formula:
I2Mn
InChi Key:
QWYFOIJABGVEFP-UHFFFAOYSA-L
SMILES:
[Mn++].[I-].[I-]
1
–
3
of
3
results
1
Manganese(II) iodide, anhydrous, 98%
CAS: 7790-33-2 MDL Number: MFCD00016225
2
Manganese(II) iodide, ultra dry, 99.99% (metals basis)
CAS: 7790-33-2 Molecular Formula: I2Mn MDL Number: MFCD00016225
3
Manganese(II) iodide, ultra dry, 99.99% (metals basis)
CAS: 7790-33-2 MDL Number: MFCD00016225