accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7789-66-4

BrSiBrBrBr

CAS RN 7789-66-4

IUPAC Name: tetrabromosilane
Synonyms: silicon tetrabromide
Molecular Weight (g/mol): 347.7
Molecular Formula: Br4Si
InChi Key: AIFMYMZGQVTROK-UHFFFAOYSA-N
SMILES: Br[Si](Br)(Br)Br
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results