accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7789-64-2

BrBrBrBrIr

CAS RN 7789-64-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 511.83
Molecular Formula: Br4Ir
InChi Key: IDTZIDJMCPQPPV-UHFFFAOYSA-N
SMILES: [Br].[Br].[Br].[Br].[Ir]
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results