accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7789-27-7

FTl

CAS RN 7789-27-7

IUPAC Name: λ¹-thallanylium fluoride
Synonyms: thallium(I) fluoride
Molecular Weight (g/mol): 223.38
Molecular Formula: FTl
InChi Key: CULOEOTWMUCRSJ-UHFFFAOYSA-M
SMILES: [F-].[Tl+]
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)