accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7787-63-5

HIBiHO

CAS RN 7787-63-5

IUPAC Name: iodobismuthanone
Synonyms: iodobismuthanone
Molecular Weight (g/mol): 353.9
Molecular Formula: BiH2IO
InChi Key: LFGCOBWXLQWPDW-UHFFFAOYSA-N
SMILES: I.O=[BiH]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results