accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7784-19-2

NH4NH4NH4FAlFFFFF

CAS RN 7784-19-2

IUPAC Name: triammonium hexafluoroalumanetriuide
Synonyms: triammonium hexafluoroalumanetriuide
Molecular Weight (g/mol): 195.09
Molecular Formula: AlF6H12N3
InChi Key: OYHBNKHFKHBTRQ-UHFFFAOYSA-K
SMILES: [NH4+].[NH4+].[NH4+].F[Al-3](F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results