accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7783-95-1

HFAgFH

CAS RN 7783-95-1

IUPAC Name: silver(II) fluoride
Synonyms: silver(II) fluoride
Molecular Weight (g/mol): 147.88
Molecular Formula: AgF2H2
InChi Key: DOVBQBITXCIRPH-UHFFFAOYSA-N
SMILES: F.F.[Ag]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)