accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7783-93-9

IUPAC Name: silver(1+) perchlorate
Synonyms: silver perchlorate
Molecular Weight (g/mol): 207.31
Molecular Formula: AgClO4
InChi Key: YDHABVNRCBNRNZ-UHFFFAOYSA-M
SMILES: [Ag+].[O-][Cl](=O)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)