accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7783-52-0

FFFIn

CAS RN 7783-52-0

IUPAC Name: indium(3+) trifluoride
Synonyms: indium(III) fluoride
Molecular Weight (g/mol): 171.81
Molecular Formula: F3In
InChi Key: JNLSTWIBJFIVHZ-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[In+3]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)