accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7782-89-0

LiNH2

CAS RN 7782-89-0

IUPAC Name: lithium(1+) azanide
Synonyms: lithium amide
Molecular Weight (g/mol): 22.96
Molecular Formula: H2LiN
InChi Key: AFRJJFRNGGLMDW-UHFFFAOYSA-N
SMILES: [Li+].[NH2-]
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (4)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)