accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CaCaCaOAsOOOOAsOOO

CAS RN 7778-44-1

IUPAC Name: tricalcium diarsorate
Synonyms: calcium arsenate arsenic acid and its salts
Molecular Weight (g/mol): 398.07
Molecular Formula: As2Ca3O8
InChi Key: RMBBSOLAGVEUSI-UHFFFAOYSA-H
SMILES: [Ca++].[Ca++].[Ca++].[O-][As]([O-])([O-])=O.[O-][As]([O-])([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)