accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CSH

CAS RN 7773-83-3

IUPAC Name: docosane-1-thiol
Synonyms: docosane-1-thiol
Molecular Weight (g/mol): 342.67
Molecular Formula: C22H46S
InChi Key: NNZMLOHQRXHPOZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCS
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)