accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 77644-45-2

H2NHNSHNNO

CAS RN 77644-45-2

IUPAC Name: 3-amino-1-[2-(morpholin-4-yl)ethyl]thiourea
Synonyms: 3-amino-1-[2-(morpholin-4-yl)ethyl]thiourea
Molecular Weight (g/mol): 204.29
Molecular Formula: C7H16N4OS
InChi Key: OBGPRENQQPWJOC-UHFFFAOYSA-N
SMILES: NNC(=S)NCCN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results