accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7758-88-5

FFFCe

CAS RN 7758-88-5

IUPAC Name: cerium(3+) trifluoride
Synonyms: cerium(3+) trifluoride
Molecular Weight (g/mol): 197.11
Molecular Formula: CeF3
InChi Key: QCCDYNYSHILRDG-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Ce+3]
Physical Form 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)