accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOHNO

CAS RN 77484-99-2

IUPAC Name: methyl 4H-furo[3,2-b]pyrrole-5-carboxylate
Synonyms: methyl 4H-furo[3,2-b]pyrrole-5-carboxylate
Molecular Weight (g/mol): 165.15
Molecular Formula: C8H7NO3
InChi Key: LIIJOCBTGRORSY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=C(N1)C=CO2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results