accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 77337-82-7

IUPAC Name: 1-bromo-2-methoxy-4-nitrobenzene
Synonyms: 1-bromo-2-methoxy-4-nitrobenzene
Molecular Weight (g/mol): 232.03
Molecular Formula: C7H6BrNO3
InChi Key: NTKADLOYTKVXQN-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1Br)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results