CAS RN 773109-13-0
IUPAC Name:
2-[2-methoxy-5-(trifluoromethoxy)phenyl]acetic acid
Synonyms:
[2-methoxy-5-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol):
250.17
Molecular Formula:
C10H9F3O4
InChi Key:
HDDOJDLMPLUGCV-UHFFFAOYSA-N
SMILES:
COC1=C(CC(O)=O)C=C(OC(F)(F)F)C=C1
1
–
1
of
1
results
1
2-Methoxy-5-(trifluoromethoxy)phenylacetic acid, 97%, Thermo Scientific™
CAS: 773109-13-0 Molecular Formula: C10H9F3O4 Molecular Weight (g/mol): 250.17 MDL Number: MFCD04972913 InChI Key: HDDOJDLMPLUGCV-UHFFFAOYSA-N Synonym: 2-methoxy-5-trifluoromethoxy phenylacetic acid,2-2-methoxy-5-trifluoromethoxy phenyl acetic acid,2-methoxy-5-trifluoromethoxy phenyl acetic acid,2-methoxy-5-trifluoromethoxyphenyl acetic acid PubChem CID: 17999353 IUPAC Name: 2-[2-methoxy-5-(trifluoromethoxy)phenyl]acetic acid SMILES: COC1=C(CC(O)=O)C=C(OC(F)(F)F)C=C1