accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7731-29-5

CH3OH

CAS RN 7731-29-5

IUPAC Name: 4-methylcyclohexan-1-ol
Synonyms: 4-methylcyclohexanol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: MQWCXKGKQLNYQG-UHFFFAOYSA-N
SMILES: CC1CCC(O)CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results