accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClFFFNH2

CAS RN 771583-81-4

IUPAC Name: 1-[4-chloro-2-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[4-chloro-2-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 209.6
Molecular Formula: C8H7ClF3N
InChi Key: VPKIGWHSTBKRTA-UHFFFAOYSA-N
SMILES: NCC1=C(C=C(Cl)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results