accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NH2FFF

CAS RN 771581-64-7

IUPAC Name: 1-[4-methyl-3-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[4-methyl-3-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 189.18
Molecular Formula: C9H10F3N
InChi Key: QPMGRSWJTRAJIK-UHFFFAOYSA-N
SMILES: CC1=C(C=C(CN)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results