accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NO

CAS RN 771573-25-2

IUPAC Name: 1-[2-(furan-2-yl)phenyl]methanamine
Synonyms: 1-[2-(furan-2-yl)phenyl]methanamine
Molecular Weight (g/mol): 173.22
Molecular Formula: C11H11NO
InChi Key: KUTREPQQCILHIM-UHFFFAOYSA-N
SMILES: NCC1=CC=CC=C1C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results